Assay Design



Gene Transcripts

Gene Symbol
FABP9
Entrez Gene ID
515385
Gene Name
fatty acid binding protein 9
Gene Aliases
-
Chromosome Location
Chr.14: 44660651 - 44664337 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192410.1 NP_001179339.1 2 - 3 273 77
GenBank mRNA CF768059.1 - 2 - 3 273 77

View Full Product Details