Assay Design



Gene Transcripts

Gene Symbol
SLC2A10
Entrez Gene ID
516135
Gene Name
solute carrier family 2 member 10
Gene Aliases
-
Chromosome Location
Chr.13: 75368042 - 75380005 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192439.3 NP_001179368.3 4 - 5 1616 99

View Full Product Details