Assay Design



Gene Transcripts

Gene Symbol
MREG
Entrez Gene ID
613445
Gene Name
melanoregulin
Gene Aliases
-
Chromosome Location
Chr.2: 103970393 - 104038722 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192953.1 NP_001179882.1 2 - 3 252 90

View Full Product Details