Assay Design



Gene Transcripts

Gene Symbol
PCDH7
Entrez Gene ID
514537
Gene Name
protocadherin 7
Gene Aliases
-
Chromosome Location
Chr.6: 49954386 - 50430389 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192975.3 NP_001179904.1 1 - 2 799 55

View Full Product Details