Assay Design



Gene Transcripts

Gene Symbol
KCNH5
Entrez Gene ID
537813
Gene Name
potassium voltage-gated channel subfamily H member 5
Gene Aliases
-
Chromosome Location
Chr.10: 74940464 - 75324314 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192989.1 NP_001179918.1 7 - 8 1371 99

View Full Product Details