Assay Design



Gene Transcripts

Gene Symbol
DYM
Entrez Gene ID
613923
Gene Name
dymeclin
Gene Aliases
-
Chromosome Location
Chr.24: 48720702 - 49123084 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192999.1 NP_001179928.1 8 - 9 1032 105

View Full Product Details