Assay Design



Gene Transcripts

Gene Symbol
BEND4
Entrez Gene ID
614525
Gene Name
BEN domain containing 4
Gene Aliases
CCDC4
Chromosome Location
Chr.6: 60982593 - 61011548 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193035.2 NP_001179964.2 4 - 5 1391 115

View Full Product Details