Assay Design



Gene Transcripts

Gene Symbol
MYMK
Entrez Gene ID
614755
Gene Name
myomaker, myoblast fusion factor
Gene Aliases
TMEM8C
Chromosome Location
Chr.11: 104396320 - 104415568 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193046.1 NP_001179975.1 1 - 2 199 82

View Full Product Details