Assay Design



Gene Transcripts

Gene Symbol
LYL1
Entrez Gene ID
615458
Gene Name
LYL1 basic helix-loop-helix family member
Gene Aliases
-
Chromosome Location
Chr.7: 12473687 - 12479882 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193074.3 NP_001180003.3 2 - 3 422 63

View Full Product Details