Assay Design



Gene Transcripts

Gene Symbol
STYX
Entrez Gene ID
616825
Gene Name
serine/threonine/tyrosine interacting protein
Gene Aliases
-
Chromosome Location
Chr.10: 11347878 - 11460270 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193128.1 NP_001180057.1 7 - 8 654 99

View Full Product Details