Assay Design



Gene Transcripts

Gene Symbol
ZKSCAN5
Entrez Gene ID
516885
Gene Name
zinc finger with KRAB and SCAN domains 5
Gene Aliases
-
Chromosome Location
Chr.25: 36890055 - 36906423 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193173.1 NP_001180102.1 2 - 3 936 117

View Full Product Details