Assay Design



Gene Transcripts

Gene Symbol
SYT17
Entrez Gene ID
532273
Gene Name
synaptotagmin 17
Gene Aliases
-
Chromosome Location
Chr.25: 16710907 - 16799124 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193210.1 NP_001180139.1 5 - 6 1069 75

View Full Product Details