Assay Design



Gene Transcripts

Gene Symbol
JDP2
Entrez Gene ID
531476
Gene Name
Jun dimerization protein 2
Gene Aliases
JUNDM2
Chromosome Location
Chr.10: 86610202 - 86655611 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193245.3 NP_001180174.1 1 - 2 177 71

View Full Product Details