Assay Design



Gene Transcripts

Gene Symbol
NLK
Entrez Gene ID
507204
Gene Name
nemo like kinase
Gene Aliases
-
Chromosome Location
Chr.19: 19651228 - 19780834 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193253.1 NP_001180182.1 7 - 8 1915 75

View Full Product Details