Assay Design



Gene Transcripts

Gene Symbol
CCDC189
Entrez Gene ID
615350
Gene Name
coiled-coil domain containing 189
Gene Aliases
C16orf93, C25H16orf93
Chromosome Location
Chr.25: 26876841 - 26882897 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195631.1 NP_001182560.1 6 - 7 729 71

View Full Product Details