Assay Design



Gene Transcripts

Gene Symbol
PYY
Entrez Gene ID
615800
Gene Name
peptide YY
Gene Aliases
-
Chromosome Location
Chr.19: 43821277 - 43822515 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040587.2 NP_001035677.1 3 - 4 359 116
GenBank mRNA BC102123.1 - 3 - 4 360 116

View Full Product Details