Assay Design



Gene Transcripts

Gene Symbol
MPV17L
Entrez Gene ID
617491
Gene Name
MPV17 mitochondrial inner membrane protein like
Gene Aliases
-
Chromosome Location
Chr.25: 13816397 - 13826839 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046602.2 NP_001040067.1 1 - 2 338 78
GenBank mRNA BC112608.1 - 1 - 2 340 78

View Full Product Details