Assay Design



Gene Transcripts

Gene Symbol
PALM2
Entrez Gene ID
100532750
Gene Name
paralemmin 2
Gene Aliases
AKAP2, PALM2-AKAP2, PALM2AKAP2
Chromosome Location
Chr.8: 99342276 - 99503025 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204119.1 NP_001191048.1 5 - 6 397 73
NM_001371509.1 NP_001358438.1 5 - 6 621 73
GenBank mRNA DN283780.1 - 4 - 5 320 73
DN511821.1 - 4 - 5 320 73
DN546048.1 - 4 - 5 320 73
EH372970.1 - 5 - 6 411 73

View Full Product Details