Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205376.2 | NP_001192305.1 | 4 - 5 | 260 | 78 |
GenBank mRNA | BC103023.1 | - | 4 - 5 | 251 | 78 |
EH182364.1 | - | 4 - 5 | 260 | 78 | |
EV683183.1 | - | 4 - 5 | 260 | 78 |