Assay Design



Gene Transcripts

Gene Symbol
KLHL42
Entrez Gene ID
781458
Gene Name
kelch like family member 42
Gene Aliases
KLHDC5
Chromosome Location
Chr.5: 82042286 - 82063595 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205415.2 NP_001192344.1 2 - 3 1187 105

View Full Product Details