Assay Design



Gene Transcripts

Gene Symbol
KCNK9
Entrez Gene ID
539586
Gene Name
potassium two pore domain channel subfamily K member 9
Gene Aliases
-
Chromosome Location
Chr.14: 3615508 - 3712832 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206824.1 NP_001193753.1 1 - 2 277 105

View Full Product Details