Product Details

SNP ID
rs74425027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112154955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCCATAAATTCTAGACTCTCTT[C/T]TACCCAAAATACAACTTTTAGAAAG
Phenotype
MIM: 612738
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC9C1 PubMed Links

Gene Details

Gene
SLC9C1
Gene Name
solute carrier family 9 member C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320531.1 Intron NP_001307460.1
NM_183061.2 Intron NP_898884.1
XM_006713603.2 Intron XP_006713666.1
XM_011512718.1 Intron XP_011511020.1
XM_011512719.1 Intron XP_011511021.1
XM_011512720.1 Intron XP_011511022.1
XM_011512721.1 Intron XP_011511023.1
XM_011512724.1 Intron XP_011511026.1
XM_011512725.2 Intron XP_011511027.1
XM_011512726.2 Intron XP_011511028.1
XM_011512727.2 Intron XP_011511029.1
XM_017006246.1 Intron XP_016861735.1
XM_017006247.1 Intron XP_016861736.1
XM_017006248.1 Intron XP_016861737.1
XM_017006249.1 Intron XP_016861738.1
XM_017006250.1 Intron XP_016861739.1

View Full Product Details