Product Details

SNP ID
rs75686093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:125084070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGCTCCTGCAGTGAGAGAGACA[C/G]AGAGGATGGTGAGAAGGACAGAACA
Phenotype
MIM: 611316
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC12A8 PubMed Links

Gene Details

Gene
SLC12A8
Gene Name
solute carrier family 12 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195483.1 Intron NP_001182412.1
NM_024628.5 Intron NP_078904.3

View Full Product Details