Product Details

SNP ID
rs77812985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129397550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTCCTCACCTGTAAAATGGAGA[C/T]GACATTGTGTGCTGAATGCCTGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EFCAB12 PubMed Links

Gene Details

Gene
EFCAB12
Gene Name
EF-hand calcium binding domain 12
There are no transcripts associated with this gene.

Gene
RPL32P3
Gene Name
ribosomal protein L32 pseudogene 3
There are no transcripts associated with this gene.

Gene
SNORA7B
Gene Name
small nucleolar RNA, H/ACA box 7B
There are no transcripts associated with this gene.

View Full Product Details