Product Details

SNP ID
rs74673718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:133576405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGAGGGAATGTGAGGTTCTTA[A/C]CATTTGGTGTTGTGGAACTGCCTGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDV3 PubMed Links

Gene Details

Gene
CDV3
Gene Name
CDV3 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134422.1 Intron NP_001127894.1
NM_001134423.2 Intron NP_001127895.1
NM_001282762.1 Intron NP_001269691.1
NM_001282763.1 Intron NP_001269692.1
NM_001282764.1 Intron NP_001269693.1
NM_001282765.1 Intron NP_001269694.1
NM_017548.4 Intron NP_060018.1
XM_005247588.4 Intron XP_005247645.1
XM_005247589.4 Intron XP_005247646.1
XM_005247590.4 Intron XP_005247647.1
XM_005247591.4 Intron XP_005247648.1
XM_005247592.4 Intron XP_005247649.1
XM_005247594.4 Intron XP_005247651.1
XM_011512961.2 Intron XP_011511263.1
XM_011512962.2 Intron XP_011511264.1
XM_017006808.1 Intron XP_016862297.1

View Full Product Details