Product Details

SNP ID
rs79260524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:135971678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATGGTTAACTTCTGTATAAAGG[A/G]TGACTTTGTGACATGAAAGCAGACC
Phenotype
MIM: 604944
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP2R3A PubMed Links

Gene Details

Gene
PPP2R3A
Gene Name
protein phosphatase 2 regulatory subunit B''alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190447.1 Intron NP_001177376.1
NM_002718.4 Intron NP_002709.2
NM_181897.2 Intron NP_871626.1
XM_006713686.3 Intron XP_006713749.1
XM_011512956.2 Intron XP_011511258.1
XM_017006787.1 Intron XP_016862276.1

View Full Product Details