Product Details

SNP ID
rs77806389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149739657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGTGGACAAATAAAAACCCTTA[C/T]AGAGGTATTTTTTATCATATGTTTT
Phenotype
MIM: 616699
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COMMD2 PubMed Links

Gene Details

Gene
COMMD2
Gene Name
COMM domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016094.3 2519 UTR 3 NP_057178.2

View Full Product Details