Product Details

SNP ID
rs79289972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:155770416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTGGCGAAGCAGAGCCAAAAAA[A/G]TCCTGTGTCAACCCCATAGTCTTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3orf33 PubMed Links

Gene Details

Gene
C3orf33
Gene Name
chromosome 3 open reading frame 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308229.1 Intron NP_001295158.1
NM_173657.2 Intron NP_775928.1
XM_005247379.4 Intron XP_005247436.1
XM_011512710.2 Intron XP_011511012.1
XM_011512711.2 Intron XP_011511013.1

View Full Product Details