Product Details

SNP ID
rs79238110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172050928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTATTGAGAGTCATGGTCTCCT[C/T]ATATCCAGGAGTGATACGAAGAATT
Phenotype
MIM: 611909
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FNDC3B PubMed Links

Gene Details

Gene
FNDC3B
Gene Name
fibronectin type III domain containing 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135095.1 Intron NP_001128567.1
NM_022763.3 Intron NP_073600.3
XM_017007062.1 Intron XP_016862551.1
XM_017007063.1 Intron XP_016862552.1
XM_017007064.1 Intron XP_016862553.1

View Full Product Details