Product Details

SNP ID
rs80236491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183255713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGAAGAGTTTTCAAAATAAAGT[A/G]CAGGCCCCATCAAGTAAAAAATGAA
Phenotype
MIM: 615333
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B3GNT5 PubMed Links

Gene Details

Gene
B3GNT5
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032047.4 761 Intron NP_114436.1
XM_005247823.4 761 Intron XP_005247880.1
XM_005247824.3 761 Intron XP_005247881.1
XM_005247825.3 761 Intron XP_005247882.1
XM_011513225.2 761 Intron XP_011511527.1
XM_011513226.2 761 Intron XP_011511528.1
XM_011513227.2 761 Intron XP_011511529.1
XM_011513228.2 761 Intron XP_011511530.1
XM_011513229.2 761 Intron XP_011511531.1
XM_017007309.1 761 UTR 5 XP_016862798.1
Gene
MCF2L2
Gene Name
MCF.2 cell line derived transforming sequence-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015078.3 761 Intron NP_055893.3
XM_011512585.2 761 Intron XP_011510887.1
XM_011512586.2 761 Intron XP_011510888.1
XM_017005943.1 761 Intron XP_016861432.1
XM_017005944.1 761 Intron XP_016861433.1
XM_017005945.1 761 Intron XP_016861434.1
XM_017005946.1 761 Intron XP_016861435.1

View Full Product Details