Product Details

SNP ID
rs75748075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183710211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGATTAGCTGGACATTTTGTTTC[A/C]AAGTTGAATTGACCATATCTAACAA
Phenotype
MIM: 613373
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
YEATS2 PubMed Links

Gene Details

Gene
YEATS2
Gene Name
YEATS domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018023.4 228 Intron NP_060493.3
XM_005247597.2 228 Intron XP_005247654.1
XM_005247598.4 228 Intron XP_005247655.1
XM_006713688.3 228 UTR 5 XP_006713751.1
XM_011512965.1 228 Intron XP_011511267.1
XM_011512966.1 228 Intron XP_011511268.1
XM_017006809.1 228 UTR 5 XP_016862298.1
XM_017006810.1 228 Intron XP_016862299.1
XM_017006811.1 228 Intron XP_016862300.1
XM_017006812.1 228 Intron XP_016862301.1
XM_017006813.1 228 Intron XP_016862302.1
XM_017006814.1 228 Intron XP_016862303.1

View Full Product Details