Product Details

SNP ID
rs79632906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:22699592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGTTCTATATAAGTTGGTGGGA[C/T]CTATTGACAGTCAGTTGTCTCACAA
Phenotype
MIM: 606619
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GBA3 PubMed Links

Gene Details

Gene
GBA3
Gene Name
glucosylceramidase beta 3 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128432.2 Intron NP_001121904.1
NM_001277225.1 Intron NP_001264154.1
NM_020973.4 Intron NP_066024.1

View Full Product Details