Product Details

SNP ID
rs79327764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17522210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTAAATGTTGATTATCCTCAATT[A/C]ATATGCAGGGATTACTATCCCCATT
Phenotype
MIM: 612676
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLRN2 PubMed Links

Gene Details

Gene
CLRN2
Gene Name
clarin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079827.2 Intron NP_001073296.1
Gene
QDPR
Gene Name
quinoid dihydropteridine reductase
There are no transcripts associated with this gene.

View Full Product Details