Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_178140.3 | Intron | NP_835260.2 | |||
XM_005248269.4 | Intron | XP_005248326.1 | |||
XM_005248271.1 | Intron | XP_005248328.1 | |||
XM_005248272.3 | Intron | XP_005248329.1 | |||
XM_006714460.2 | Intron | XP_006714523.1 | |||
XM_011513992.2 | Intron | XP_011512294.1 | |||
XM_011513993.2 | Intron | XP_011512295.1 | |||
XM_011513994.2 | Intron | XP_011512296.1 | |||
XM_011513995.2 | Intron | XP_011512297.1 | |||
XM_011513996.2 | Intron | XP_011512298.1 | |||
XM_017009245.1 | Intron | XP_016864734.1 | |||
XM_017009246.1 | Intron | XP_016864735.1 |