Product Details

SNP ID
rs79045306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:41310204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCCTCATTCTCTGCATTTACAT[G/T]ATTGTGTGGTAAATAGGGAGACATA
Phenotype
MIM: 617016
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLCXD3 PubMed Links

Gene Details

Gene
PLCXD3
Gene Name
phosphatidylinositol specific phospholipase C X domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005473.2 4481 UTR 3 NP_001005473.1
XM_017009436.1 4481 Intron XP_016864925.1
XM_017009437.1 4481 Intron XP_016864926.1
XM_017009438.1 4481 Intron XP_016864927.1

View Full Product Details