Product Details
- SNP ID
-
rs77879475
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:71719387 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGTACCCGTGCCCAGGAGGACGCC[A/G]AGCTCCAGCCCCGAGCCCTGGACAT
- Phenotype
-
MIM: 602606
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CARTPT
PubMed Links
Gene Details
- Gene
- CARTPT
- Gene Name
- CART prepropeptide
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004291.3 |
225 |
Missense Mutation |
AAG,GAG |
K32E |
NP_004282.1 |
View Full Product Details