Product Details

SNP ID
rs75770132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:75372533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCAAATTATGAACAGCATTTTTC[A/T]ATAAGGCATATTCCATGCACATCAT
Phenotype
MIM: 604677
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
COL4A3BP PubMed Links

Gene Details

Gene
COL4A3BP
Gene Name
collagen type IV alpha 3 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130105.1 4064 UTR 3 NP_001123577.1
NM_005713.2 4064 UTR 3 NP_005704.1
NM_031361.2 4064 UTR 3 NP_112729.1
XM_006714513.2 4064 Intron XP_006714576.1
XM_011543090.2 4064 UTR 3 XP_011541392.1
XM_011543091.2 4064 UTR 3 XP_011541393.1
XM_017008919.1 4064 Intron XP_016864408.1

View Full Product Details