Product Details

SNP ID
rs76690871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:75611835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCACTAAACTGCTGCCTGACTAA[C/T]GAACTGAACTGACTATGCTATCATC
Phenotype
MIM: 605003
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SENP6 PubMed Links

Gene Details

Gene
SENP6
Gene Name
SUMO1/sentrin specific peptidase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100409.2 Intron NP_001093879.1
NM_001304792.1 Intron NP_001291721.1
NM_015571.3 Intron NP_056386.2
XM_006715457.3 Intron XP_006715520.1
XM_011535746.2 Intron XP_011534048.1
XM_011535747.2 Intron XP_011534049.1
XM_017010719.1 Intron XP_016866208.1
XM_017010720.1 Intron XP_016866209.1
XM_017010721.1 Intron XP_016866210.1
XM_017010722.1 Intron XP_016866211.1
XM_017010723.1 Intron XP_016866212.1
XM_017010724.1 Intron XP_016866213.1
XM_017010725.1 Intron XP_016866214.1

View Full Product Details