Product Details

SNP ID
rs75206923
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27140020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGATGGGCTCCACTAAATGCTA[A/G]AACCTCCAGGAAAACTTCGTGGTGG
Phenotype
MIM: 615012 MIM: 615013 MIM: 615044 MIM: 615045 MIM: 602833
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HIST1H2AG PubMed Links

Gene Details

Gene
HIST1H2AG
Gene Name
histone cluster 1, H2ag
There are no transcripts associated with this gene.

Gene
HIST1H2AH
Gene Name
histone cluster 1, H2ah
There are no transcripts associated with this gene.

Gene
HIST1H2BJ
Gene Name
histone cluster 1, H2bj
There are no transcripts associated with this gene.

Gene
HIST1H2BK
Gene Name
histone cluster 1, H2bk
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001312653.1 Intron NP_001299582.1
NM_080593.2 Intron NP_542160.1
Gene
HIST1H4I
Gene Name
histone cluster 1, H4i
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003495.2 Intron NP_003486.1
Gene
MIR3143
Gene Name
microRNA 3143
There are no transcripts associated with this gene.

View Full Product Details