Product Details

SNP ID
rs78945358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32841717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGCCGAGTCCCATGTTCATCCA[C/T]GTAGTAGAGTCCAGGACCCTATAAG
Phenotype
MIM: 177046 MIM: 170260 MIM: 170261
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMB8 PubMed Links

Gene Details

Gene
PSMB8
Gene Name
proteasome subunit beta 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004159.4 1066 Missense Mutation ATG,GTG M182V NP_004150.1
NM_148919.3 1066 Missense Mutation ATG,GTG M186V NP_683720.2
Gene
PSMB8-AS1
Gene Name
PSMB8 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
TAP1
Gene Name
transporter 1, ATP binding cassette subfamily B member
There are no transcripts associated with this gene.

Gene
TAP2
Gene Name
transporter 2, ATP binding cassette subfamily B member
There are no transcripts associated with this gene.

View Full Product Details