Product Details
- SNP ID
-
rs75559015
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:43334237 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGCCCTGTGAATATAATATGGCTT[G/T]TAAAGCAGGAAGCCCCCCATTTGCA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF318
PubMed Links
Gene Details
- Gene
- ZNF318
- Gene Name
- zinc finger protein 318
View Full Product Details