Product Details

SNP ID
rs74535144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150389496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGTCCAAGGAGCCAGGTGGTG[A/G]CAGGAGCACCCTAGCTGCATGTGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF775 PubMed Links

Gene Details

Gene
ZNF775
Gene Name
zinc finger protein 775
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173680.3 Intron NP_775951.2

View Full Product Details