Product Details

SNP ID
rs79281353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150627452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGTAACTCATGAAAGGCAATGC[A/C]ATGAGATAGAAGGTCCAATAGGAAG
Phenotype
MIM: 616960
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GIMAP6 PubMed Links

Gene Details

Gene
GIMAP6
Gene Name
GTPase, IMAP family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244071.1 1402 UTR 3 NP_001231000.1
NM_001244072.1 1402 UTR 3 NP_001231001.1
NM_024711.5 1402 UTR 3 NP_078987.3

View Full Product Details