Product Details

SNP ID
rs77293332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:65960725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGCCAACTCTTTATTTCCATAA[C/T]AGAAAATCTTTTATTTCCACCTTTA
Phenotype
MIM: 611499 MIM: 608838
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUSB PubMed Links

Gene Details

Gene
GUSB
Gene Name
glucuronidase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000181.3 2003 UTR 3 NP_000172.2
NM_001284290.1 2003 UTR 3 NP_001271219.1
NM_001293104.1 2003 UTR 3 NP_001280033.1
NM_001293105.1 2003 UTR 3 NP_001280034.1
XM_005250297.3 2003 UTR 3 XP_005250354.1
XM_011516113.1 2003 UTR 3 XP_011514415.1
XM_011516114.1 2003 UTR 3 XP_011514416.1
XM_017012091.1 2003 UTR 3 XP_016867580.1
XM_017012092.1 2003 UTR 3 XP_016867581.1
XM_017012093.1 2003 UTR 3 XP_016867582.1
Gene
VKORC1L1
Gene Name
vitamin K epoxide reductase complex subunit 1 like 1
There are no transcripts associated with this gene.

View Full Product Details