Product Details
- SNP ID
-
rs75829477
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:86053020 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGCTTTAAATGATTCAAAGGAATT[A/C]GGAGGGATGGTATCTTACATGCTGT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PSKH2
PubMed Links
Gene Details
- Gene
- PSKH2
- Gene Name
- protein serine kinase H2
View Full Product Details