Product Details

SNP ID
rs76178785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123320759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTTAAGTAAGTTTCTTGTCAGA[C/T]ACAATAAACTATTTTTAGGAAGGAT
Phenotype
MIM: 611941
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATAD2 PubMed Links

Gene Details

Gene
ATAD2
Gene Name
ATPase family, AAA domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014109.3 3076 UTR 3 NP_054828.2
XM_011516994.2 3076 UTR 3 XP_011515296.1
XM_011516995.2 3076 Intron XP_011515297.1
XM_011516996.2 3076 UTR 3 XP_011515298.1

View Full Product Details