Product Details

SNP ID
rs76597597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:93922033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTAGGAATCCCAGTCAGAAGT[C/T]CCAGCCTGCCACTGTTCTCTGATGC
Phenotype
MIM: 605993
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR378D2 PubMed Links

Gene Details

Gene
MIR378D2
Gene Name
microRNA 378d-2
There are no transcripts associated with this gene.

Gene
PDP1
Gene Name
pyruvate dehyrogenase phosphatase catalytic subunit 1
There are no transcripts associated with this gene.

View Full Product Details