Product Details

SNP ID
rs76011963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:71843161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCGGGAGAGCTGGCAGATCAGAG[A/G]GGGACTTTCCAGATAAATCCCATCT
Phenotype
MIM: 603628
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MSC PubMed Links

Gene Details

Gene
MSC
Gene Name
musculin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005098.3 Intron NP_005089.2
Gene
MSC-AS1
Gene Name
MSC antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details