Product Details

SNP ID
rs77689867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:73423498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCCAAGCCCGACACTGCTGAGT[A/G]AGATTGGAAGGGCTCTGCTGAGTCT
Phenotype
MIM: 605920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STAU2 PubMed Links

Gene Details

Gene
STAU2
Gene Name
staufen double-stranded RNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164380.1 Intron NP_001157852.1
NM_001164381.1 Intron NP_001157853.1
NM_001164382.1 Intron NP_001157854.1
NM_001164383.1 Intron NP_001157855.1
NM_001164384.1 Intron NP_001157856.1
NM_001164385.1 Intron NP_001157857.1
NM_014393.2 Intron NP_055208.2
Gene
STAU2-AS1
Gene Name
STAU2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details