Product Details

SNP ID
rs75896465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:4991044 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGTTTTTTTCTGAATGATAGAA[A/G]GTTAGACAAGCCATCTCTGGGTGCT
Phenotype
MIM: 606414 MIM: 147796
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INSL6 PubMed Links

Gene Details

Gene
INSL6
Gene Name
insulin like 6
There are no transcripts associated with this gene.

Gene
JAK2
Gene Name
Janus kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322194.1 Intron NP_001309123.1
NM_001322195.1 Intron NP_001309124.1
NM_001322196.1 Intron NP_001309125.1
NM_001322198.1 Intron NP_001309127.1
NM_001322199.1 Intron NP_001309128.1
NM_001322204.1 Intron NP_001309133.1
NM_004972.3 Intron NP_004963.1

View Full Product Details